LIQUITHIEF 1000 MM PP BX20
Catalog # 77576-869
Supplier: SAMPLING SYSTEMS LTD
New Product
LIQUITHIEF 1000 MM PP BX20
Catalog # 77576-869
Supplier: SAMPLING SYSTEMS LTD
Supplier Number: 8050P-1000
New Product
Frequently Bought Together
High Vacuum Grease, DuPont
Supplier: Liveo
Sterile Swabs, MICROLOGY INC
Supplier: MICROLOGY INC
BD Trypticase™ Soy Agar, BD Diagnostics
Supplier: BD
SpeedStreaks™ Loops and Needles in Rigid Containers, Hardy Diagnostics
Supplier: Hardy Diagnostics
SterEM™ Tryptic Soy Agar (TSA), Irradiated
Supplier: Hardy Diagnostics
BioBall® SingleShot 30
Supplier: bioMerieux
Tryptic Soy Broth (TSB)
Supplier: Hardy Diagnostics
MAGNA AMP LOG 6 PK50
Supplier: MESA LABS MS
EnviroTrans™, Saline 0.85%, 0.5 mL Swab, Hardy Diagnostics
Supplier: Hardy Diagnostics
INDICATOR CHEM STEAM PK=BX250
Supplier: Steris
Lok-Tight™ Contact Plates, Hardy Diagnostics
Supplier: Hardy Diagnostics
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
AnaeroGen™ by Oxoid AnaeroGen Anaerobic Gas Generator, Hardy Diagnostics
Supplier: Hardy Diagnostics
Non Irradiated Media 100 mm Plates, BioMerieux
Supplier: bioMerieux