Order Entry
United States
Orders LinkContactUsLinkComponent
Thymol blue indicator

Thymol blue indicator

Glass Bottle, 25 g
Catalog # 77873-192
Irritant
Supplier:   Ambeed
Supplier #:  A750751-25G
Thymol blue indicator
Catalog # 77873-192
Supplier:  Ambeed
CAS Number:  76-61-9
Thymol blue indicator

Glass Bottle, 25 g

CAS Number:  76-61-9
Price
$19.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: for Research Use Only

Synonyms:  Thymolsulphonphthalein, Thymolsulfonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
Specifications

Specifications

  • Pack type:
    Glass Bottle
  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.