ANTIBODY IL-10R A-3 IGG1 200UG/ML
Catalog # MSPP-SC-365374
Supplier: SANTA CRUZ BIOTECHNOLOGY MS
ANTIBODY IL-10R A-3 IGG1 200UG/ML
Catalog # MSPP-SC-365374
Supplier: SANTA CRUZ BIOTECHNOLOGY MS
Supplier Number: SC-365374
Frequently Bought Together
Immobiline™ DryStrip Gels, Cytiva
Supplier: Cytiva
Protein Molecular Weight Marker Mix, Rockland Immunchemicals
Supplier: Rockland Immunochemical
Deoxynucleotides (dNTPs), G-Biosciences
Supplier: G-Biosciences
Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99.9% for electrophoresis
Supplier: G-Biosciences
OSM (227AA) HUMAN >95% 50UG
Supplier: Genscript
MOPS, Free Acid, Molecular Biology Grade, EMD Millipore
Supplier: MilliporeSigma
IPG Buffers, Cytiva
Supplier: Cytiva
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×4 Configuration
Supplier: Alkali Scientific
BUFFER A.C.E. PHOSPHATE 50MM PH 6.5
Supplier: 0000014548
Buffer P2 (Green), Zymo Research
Supplier: Zymo Research
Sapphire Prestained Protein Standard 10 to 180 kDa, Rockland Immunochemicals
Supplier: Rockland Immunochemical
PEPTIDE ALFA ELUTION 2MG
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS
RNA RECOVERY BUFFER 10 ML
Supplier: Zymo Research
Pierce™ Ni-NTA Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
ScanLater™ Western Blot Assay Kit, Molecular Devices
Supplier: Molecular Devices
Antibody Fab Fragmentation, G-Biosciences
Supplier: G-Biosciences
GelBond® Film
Supplier: Lonza