ACTIVITY KIT P450 DEMETHYLATION 2 PLATES
Catalog # 104090-612
Supplier: ARBOR ASSAYS MS
ACTIVITY KIT P450 DEMETHYLATION 2 PLATES
Catalog # 104090-612
Supplier: ARBOR ASSAYS MS
Supplier Number: K011-F1
Frequently Bought Together
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MARKER MAURICE CIEF PI-8.40 LYOPH 210UL
Supplier: 0000014126
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences