APC CYANINE7 ANHU CD335 (NKP46) 100TEST
Catalog # 77781-858
Supplier: Biolegend
New Product
APC CYANINE7 ANHU CD335 (NKP46) 100TEST
Catalog # 77781-858
Supplier: Biolegend
Supplier Number: 331950
New Product
Frequently Bought Together
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
100 0544 ERASTIN
Supplier: STEMCELL Technologies
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
Supplier: STEMCELL Technologies
KIT PCR ASSAY ELISA HU PAPPA -20C 2PLATE
Supplier: RAYBIOTECH LIFE INC MS
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
KIT BD-TAU ADVANTAGE PLUS
Supplier: QUANTERIX CORPORATION MS
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
100 0249 FORSKOLIN 25MG
Supplier: STEMCELL Technologies
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences