PE ANTI-MOUSE KLRG1 (MAFA) RECOM 25UG
Catalog # 77780-660
Supplier: Biolegend
New Product
PE ANTI-MOUSE KLRG1 (MAFA) RECOM 25UG
Catalog # 77780-660
Supplier: Biolegend
Supplier Number: 164903
New Product
Frequently Bought Together
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
Supplier: Revvity Health Sciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
100 0544 ERASTIN
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio