LPX SARS-COV-2 SEROLOGICAL PANEL STD 1VL
Catalog # 77759-520
Supplier: Biolegend
New Product
LPX SARS-COV-2 SEROLOGICAL PANEL STD 1VL
Catalog # 77759-520
Supplier: Biolegend
Supplier Number: 741134
New Product
Frequently Bought Together
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
73254 METFORMIN HYDROCHLORIDE 5G
Supplier: STEMCELL Technologies
72262 ALL TRANS RETINOIC ACID 50MG
Supplier: STEMCELL Technologies
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
Supplier: Revvity Health Sciences
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics