SCANNER BARCODE 2D WDI4700 W/USB CABLE
Catalog # MSPP-6041734
Supplier: CDW COMPUTER CENTERS MS
SCANNER BARCODE 2D WDI4700 W/USB CABLE
Catalog # MSPP-6041734
Supplier: CDW COMPUTER CENTERS MS
Supplier Number: 6041734
Frequently Bought Together
SP Bel-Art Clavies® Biohazard Bag Holders, Autoclavable, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
VWR® Standard Dry Block Heaters, 120 V
Supplier: VWR International
Cartridges for Water Purification Systems
Supplier: ELGA LabWater
Accessories for Forma™ Steri-Cycle™ CO₂ Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
Accessories for Forma™ Series II 3110 Water-Jacketed CO₂ Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Supplier: VWR International
VWR® Advanced Mini Dry Block Heater and Mini Dry Block Heater with Heated Lid, 120 V
Supplier: VWR International
VWR® Advanced Dry Block Heaters
Supplier: VWR International
SUBSCRIPTION PLUS SMS ENTERPRISE
Supplier: Stirling Ultracold
Branson SFX150 Sonifier® Cell Disruptor with Handheld Converter and 1/8" Microtip, 120V
Supplier: Branson Ultrasonics
Pitched Blade Propellers, with Set Screws, Caframo
Supplier: Caframo
Arduino Uno Development Board, Mouser Electronics
Supplier: 0000014675
Pre-Treatment Kits and Cartridges for Water Purification Systems
Supplier: ELGA LabWater
Fast-Freeze® Flasks and Adapters, Labconco®
Supplier: Labconco
Accessories for Rotary Vane and Hybrid Vacuum Pumps, Labconco
Supplier: Labconco
Accessories for AriumPro Type 1 Water Systems
Supplier: Sartorius
Saunders® Aseptic Diaphragm Valves, Holland Applied Technologies
Supplier: Holland Applied Technologies
Stainless Steel Container for Waring® Blenders, 360 ml, Eberbach
Supplier: Eberbach
CALIPER ELCTRNIC IP67-PLUS 0-4IN/0-100MM
Supplier: FOWLER HIGH PRECISION MS