ANTIBODY PURIFIED ANTI-TAU 316-355 25UL
77760-614
New Product
Frequently Bought Together
Canine Plasma, Equitech- Bio, Inc.
: Equitech-Bio
100 1042 CHIR99021
: STEMCELL Technologies
72192 PROSTAGLANDIN E2 1MG
: STEMCELL Technologies
100 0251 ELTROMBOPAG 5MG
: STEMCELL Technologies
BLOCKER AQB013 AQUAPORIN 5MG
: FOCUS BIOMOLECULES MS
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
: Innovative Research Inc
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
: Equitech-Bio
72262 ALL TRANS RETINOIC ACID 50MG
: STEMCELL Technologies
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
: INDIGO BIOSCIENCES INC MS
73254 METFORMIN HYDROCHLORIDE 5G
: STEMCELL Technologies
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
: Invitrogen
AGONIST RESINIFERATOXIN TRPV1 100UG
: FOCUS BIOMOLECULES MS