CELL RAW-ASC KO-GSDMD 3-7X10E6 CELLS
Catalog # MSPP-RAWKOGSDM
Supplier: Invivogen
CELL RAW-ASC KO-GSDMD 3-7X10E6 CELLS
Catalog # MSPP-RAWKOGSDM
Supplier: Invivogen
Supplier Number: RAW-KOGSDMD
Frequently Bought Together
VWR® Acrylamide and Bis-Acrylamide
Supplier: VWR
Mix & Go! 2X Stock Wash Buffer, Zymo Research
Supplier: Zymo Research
Pre-Cut Western Blotting Membranes, Azure Biosystems
Supplier: Azure Biosystems
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×4 Configuration
Supplier: Alkali Scientific
OSM (227AA) HUMAN >95% 50UG
Supplier: Genscript
Antibody Fab Fragmentation, G-Biosciences
Supplier: G-Biosciences
Urea ≥99%, white prills, Ultrapure
Supplier: MP Biomedicals
RNA RECOVERY BUFFER 10 ML
Supplier: Zymo Research
Pierce™ Ni-NTA Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
DNA Elution Buffer, Zymo Research
Supplier: Zymo Research
Deoxynucleotides (dNTPs), G-Biosciences
Supplier: G-Biosciences
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
KIT DENSITY MARKER BEADS 6 VIALS POLYETH
Supplier: 0000021067
PINPOINT BINDING BUFFER 6 ML F/ISOLATION
Supplier: Zymo Research
Sapphire Prestained Protein Standard 10 to 180 kDa, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Buffer P1 (Red), Zymo Research
Supplier: Zymo Research
g-DNA Wash Buffer, Zymo Research
Supplier: Zymo Research
GelBond® Film
Supplier: Lonza