Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant FGF-8 (from E. coli)

Human Recombinant FGF-8 (from E. coli)

Vial, 100 µg
Catalog # 77753-460
Supplier:   VWR International
Human Recombinant FGF-8 (from E. coli)
Catalog # 77753-460
Supplier:  VWR International
CAS Number:  
Human Recombinant FGF-8 (from E. coli)
Human Recombinant FGF-8 (from E. coli)

Vial, 100 µg

CAS Number:  
Price
$410.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Fibroblast growth factor 8 (FGF-8) is a critical mitogenic factor that is required for normal development of the eye, ear, brain, and limb. FGF-8 functions broadly to promote cell proliferation, differentiation, and migration. Overexpression of FGF-8 increases tumor growth and angiogenesis. Human and mouse FGF-8 proteins show 100% homology.

  • High quality
  • Low endotoxin
  • Affordable

Caution: For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    100 µg
  • Storage Conditions:
    At or below –20 °C
  • Molecular Weight:
    22.5 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity assay: NR6R 3T3 cell proliferation; ED50 ≤150 ng/ml; Bioactivity ≥6.7×10³ units/mg
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P55075/P37237
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Protein/Peptide Name:
    FGF-8
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Tested Applications:
    Bioassay
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR
  • Cat. No.:
    77753-460
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    Androgen-induced growth factor|AIGF|Heparin-binding growth factor 8|HBGF-8|FGF-8b isoform
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.