Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant IL-17A (from E. coli)

Human Recombinant IL-17A (from E. coli)

Vial, 1 mg
Catalog # 77751-816
Supplier:   VWR International
Human Recombinant IL-17A (from E. coli)
Catalog # 77751-816
Supplier:  VWR International
CAS Number:  
Human Recombinant IL-17A (from E. coli)
Human Recombinant IL-17A (from E. coli)

Vial, 1 mg

CAS Number:  
Price
$2,940.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Interleukin 17A (IL-17A), also known as CTLA-8, is a member of the IL-17 family of proteins. IL-17A is a proinflammatory cytokine that is secreted by activated CD4+ and CD8+ T lymphocytes. IL-17A acts through its receptor, IL-17R, to promote increased cytokine and chemokine secretion. In turn, the cytokines and chemokines mediate the immunoregulatory function of IL-17A by promoting the proliferation, maturation, and chemoattraction of neutrophils to inflammatory sites.

  • High quality
  • Low endotoxin
  • Affordable

Elevated levels of IL-17A are associated with rheumatoid arthritis, airway inflammation, allograft rejection, inflammatory bowel disease, psoriasis, cancer, and multiple sclerosis. Human, mouse, and rat IL-17A show activity on mouse cells.

Caution: For research use only.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    15.7/31.3 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity assay: Production of IL-6 from mouse 3T3 cells; ED50 ≤10 ng/ml; Bioactivity ≥1.0×10⁵ units/mg
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    Q16552
  • Formulation:
    0.1% trifluoroacetic acid (TFA)
  • Protein/Peptide Name:
    IL-17A
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Tested Applications:
    Bioassay
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    MIVKAGITIP RNPGCPNSED KNFPRTVMVN LNIHNRNTNT NPKRSSDYYN RSTSPWNLHR NEDPERYPSV IWEAKCRHLG CINADGNVDY HMNSVPIQQE ILVLRREPPH CPNSFRLEKI LVSVGCTCVT PIVHHVA
  • Cat. No.:
    77751-816
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    IL-17|CTLA-8
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.