Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant FGF-6 (from E. coli)

Human Recombinant FGF-6 (from E. coli)

Vial, 500 µg
Catalog # 77751-012
Supplier:   VWR International
Human Recombinant FGF-6 (from E. coli)
Catalog # 77751-012
Supplier:  VWR International
CAS Number:  
Human Recombinant FGF-6 (from E. coli)
Human Recombinant FGF-6 (from E. coli)

Vial, 500 µg

CAS Number:  
Price
$1,960.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Fibroblast growth factor 6 (FGF-6) is a heparin-binding growth factor that is expressed in epithelial and mesenchymal lineages. FGF-6 binds and signals through the FGF receptors FGFR1, FGFR2, and FGFR4. FGF-6 functions as a mitogen for vascular endothelial cells and fibroblasts. FGF-6 is also an important factor driving muscle differentiation and regeneration.

  • High quality
  • Low endotoxin
  • Affordable

Caution: For research use only.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    500 µg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    18.9 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P48061
  • Formulation:
    10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5
  • Protein/Peptide Name:
    FGF-6
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
  • Cat. No.:
    77751-012
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    Heparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.