Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant FGF-6 (from E. coli)

Human Recombinant FGF-6 (from E. coli)

Vial, 25 µg
Catalog # 77751-010
Supplier:   VWR International
Human Recombinant FGF-6 (from E. coli)
Catalog # 77751-010
Supplier:  VWR International
CAS Number:  
Human Recombinant FGF-6 (from E. coli)
Human Recombinant FGF-6 (from E. coli)

Vial, 25 µg

CAS Number:  
Price
$163.33
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Fibroblast growth factor 6 (FGF-6) is a heparin-binding growth factor that is expressed in epithelial and mesenchymal lineages. FGF-6 binds and signals through the FGF receptors FGFR1, FGFR2, and FGFR4. FGF-6 functions as a mitogen for vascular endothelial cells and fibroblasts. FGF-6 is also an important factor driving muscle differentiation and regeneration.

  • High quality
  • Low endotoxin
  • Affordable

Caution: For research use only.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    25 µg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    18.9 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P48061
  • Formulation:
    10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5
  • Protein/Peptide Name:
    FGF-6
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    MGTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
  • Cat. No.:
    77751-010
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    Heparin secretory-transforming protein 2|HST-2|HSTF-2|Heparin-binding growth factor 6|HBGF-6
  • Conjugation:
    Unconjugated

Specifications