Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant PDGF-AB (from E. coli)

Human Recombinant PDGF-AB (from E. coli)

Vial, 500 µg
Catalog # 77750-748
Supplier:   VWR International
Human Recombinant PDGF-AB (from E. coli)
Catalog # 77750-748
Supplier:  VWR International
CAS Number:  
Human Recombinant PDGF-AB (from E. coli)
Human Recombinant PDGF-AB (from E. coli)

Vial, 500 µg

CAS Number:  
Price
$2,286.67
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells. PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues.

  • High quality
  • Low endotoxin
  • Affordable

PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.

Caution: For research use only.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    500 µg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    Alpha:14.4 kDa; Beta: 12.4 kDa; Total 26.8 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin Level:
    Low
  • Purity:
    ≥95%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Biological Activity:
    Bioactivity assay: NR6R-3T3 cell proliferation; ED50 ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    P04085/P01127
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Protein/Peptide Name:
    PDGF-AB
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Cytokine
  • Animal-Free:
    N
  • Tested Applications:
    Bioassay
  • Grade:
    RUO
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Sequence:
    Alpha chain: MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT Beta chain: MSLGSLTIAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRPTQVQLRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETVAAARPVT
  • Cat. No.:
    77750-748
  • Shipping Temperature:
    RT
  • Protein Synonyms:
    GDGF|ODGF
  • Conjugation:
    Unconjugated

Specifications