BEAD STREP-TACTIN XT MAGSTREP 5% 30UM
Catalog # MSPP-2-5090010
Supplier: IBA MS
BEAD STREP-TACTIN XT MAGSTREP 5% 30UM
Catalog # MSPP-2-5090010
Supplier: IBA MS
Supplier Number: 2-5090-010
Frequently Bought Together
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
MARKER MAURICE CIEF PI-8.40 LYOPH 210UL
Supplier: PROTEIN SIMPLE MS
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Sera-Mag SpeedBeads® Neutravidin Magnetic Beads, Cytiva
Supplier: Cytiva
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Western Blot Boxes
Supplier: Alkali Scientific