ESF 921 DELTA SERIES METHIONINE DEF 1L
Catalog # 77705-796
Supplier: EXPRESSION SYSTEMS LLC
New Product
ESF 921 DELTA SERIES METHIONINE DEF 1L
Catalog # 77705-796
Supplier: EXPRESSION SYSTEMS LLC
Supplier Number: 96-200-01
New Product
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
VWR® TBS Buffer, 20X Liquid
Supplier: VWR
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
VWR® Disposable Petri Dishes
Supplier: VWR International
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
AMMONIUM CITRATE DIBASIC 2.5 M 200ML
Supplier: HAMPTON RESEARCH MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Magnesium sulfate 1 M in aqueous solution
Supplier: G-Biosciences
Research Buffers and Reagents, RNase and DNase free, G-Biosciences
Supplier: G-Biosciences
Sodium chloride 0.9% in water normal saline solution
Supplier: G-Biosciences
HisTrap™ Excel Affinity Chromatography Columns
Supplier: Cytiva
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek