Orders LinkContactUsLinkComponent
Sterilisation Baskets, Side Perforated, Flat Base
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 480×480×100 mm
  77666-194
Supplier #:  WP-4572
Sterilisation Baskets, Side Perforated, Flat Base
  77666-194
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 480×480×100 mm
$213.00
Each
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
  • Dimensions:
    480×480×100 mm
  • Material:
    Stainless steel 304