Orders LinkContactUsLinkComponent
Sterilisation Baskets, Side Perforated, Flat Base
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 480×360×70 mm
Catalog # 77666-184
Supplier #:  WP-4567
Sterilisation Baskets, Side Perforated, Flat Base
Catalog # 77666-184
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 480×360×70 mm
Price
$188.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
Specifications
  • Dimensions:
    480×360×70 mm
  • Material:
    Stainless steel 304