Iris Scissors, Supercut
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
- Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
- Sharp tips
Specifications
- Description:Iris scissors
- Length:11.5 cm (4.5")
- Material:Stainless steel
- Curvature:Straight