Orders LinkContactUsLinkComponent
Iris Scissors, Supercut
Iris Scissors, Supercut
Iris scissors, 11.5 cm (4.5"), German steel, Straight
Catalog # 77661-872
Supplier #:  14225-G
Iris Scissors, Supercut
Catalog # 77661-872
Iris Scissors, Supercut
Iris scissors, 11.5 cm (4.5"), German steel, Straight
Price
$108.01
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
  • Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
  • Sharp tips
Specifications
  • Description:
    Iris scissors
  • Length:
    11.5 cm (4.5")
  • Material:
    German steel
  • Curvature:
    Straight