Orders LinkContactUsLinkComponent
Iris Scissors, Supercut
Iris scissors, 12.5 cm (5.0"), German steel, Curved
  77661-868
Iris Scissors, Supercut
  77661-868
Iris Scissors, Supercut
Iris scissors, 12.5 cm (5.0"), German steel, Curved
$106.01
Each
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
  • Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
  • Sharp tips
  • Description:
    Iris scissors
  • Length:
    12.5 cm (5.0")
  • Material:
    German steel
  • Curvature:
    Curved