Orders LinkContactUsLinkComponent
Ring Handled and Angled Operating Scissors
Ring Handled and Angled Operating Scissors
Ring-handled and angled operating scissors, german, 14 cm, Premium german steel
Catalog # 77662-116
Supplier #:  500047-G
Ring Handled and Angled Operating Scissors
Catalog # 77662-116
Ring Handled and Angled Operating Scissors
Ring-handled and angled operating scissors, german, 14 cm, Premium german steel
Price
$157.01
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ring-handled and angled operating scissors are a pair of surgical scissors commonly used in general surgery for cutting and dissecting tissues.
  • Angled scissors for greater flexibility and control in confined spaces
  • Sharp/sharp tips
The 14 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. The angle of the blades allows for greater flexibility and precision in cutting in areas that may be hard to reach with straight scissors.
These surgical scissors have two sharp, pointed tips. They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved.
Specifications
  • Description:
    Ring-handled and angled operating scissors, german
  • Length:
    14 cm
  • Material:
    Premium german steel