Orders LinkContactUsLinkComponent
Spring Scissors, Round Handles
Spring Scissors, Round Handles
6.5 mm, Straight german, Round
Catalog # 77661-734
Supplier #:  14111-G
Spring Scissors, Round Handles
Catalog # 77661-734
Spring Scissors, Round Handles
6.5 mm, Straight german, Round
Price
$199.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Round handle spring scissors have a rounded, ergonomic handle design and a spring mechanism that allows for smooth, precise cutting action. The rounded handle design offers a comfortable and natural grip, reducing hand fatigue and improving control during use. This can be especially beneficial for long procedures that require a lot of cutting. The spring mechanism provides a smooth, precise cutting action, which reduces the risk of tissue damage or trauma.
  • 14 cm (5.5") long
  • 6.5 mm blades
  • Stainless steel
The 6.5 mm curved blade is often used in ophthalmology to make precise incisions in the cornea or delicate tissues. The straight blades are more often used in microdissection, microsurgery or plastic surgery. These stainless steel spring scissors are durable and may be sterilized by chemical sterilant or autoclaving.
Specifications
  • Blade length:
    6.5 mm
  • Blade type:
    Straight german
  • Handle:
    Round