Order Entry
United States
Orders LinkContactUsLinkComponent
Gradient Makers

Gradient Makers

Gradient maker, 100 ml total volume
Catalog # MSPP-SG100
Marketsource
Supplier:   HOEFER INC MS
Supplier #:  SG100
Gradient Makers
Catalog # MSPP-SG100
Supplier:  HOEFER INC MS
Marketsource
Gradient Makers
Gradient Makers

Gradient maker, 100 ml total volume

Price
$650.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Ideal for generating gradients of polyacrylamide, sucrose and cesiium chloride.

  • Milled of heavy acrylic plastic with high quality, leak-free Teflon® valves
  • Flat base provides stability on magnetic stir
  • Comes with a support rod for attaching to a ring stand
  • Takes Luer or 4 mm int.Ø tubing
  • With included adapters, can also take 2 mm int.Ø or 22-gauge tubing

The 500 ml SG500 Gradient Maker is made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base. An easy-to-use push-pull valve opens and closes the passage between the reservoir and mixing chambers. Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns. Ideal for use with the SE615 Multiple Gel Caster.

Ordering information: SG15, SG30, SG50 and SG100 include Gradient Maker with Teflon Valves; Adapter Barbed Fitting; Luer Outlet Fitting; 22-gauge Needle; Support Rod. SG500 includes Gradient Maker with Push-Pull Valve; Hose Clamps (4 pieces).

Specifications
  • Description:
    Gradient maker, 100 ml total volume
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.