RESIN STREP TACTIN XT 4FLOW 50% 20ML
Catalog # MSPP-2-5010010
Supplier: IBA MS
RESIN STREP TACTIN XT 4FLOW 50% 20ML
Catalog # MSPP-2-5010010
Supplier: IBA MS
Supplier Number: 2-5010-010
Frequently Bought Together
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Disposable Serological Pipettes
Supplier: VWR International
Eppendorf® Flex-Tubes® Microtubes, 1.5 ml
Supplier: Eppendorf
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
BUFFER TBS BLOCKING READY-TO-USE 500ML
Supplier: Li-Cor
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
Plasmid+® Media, Thomson Instrument Company
Supplier: THOMSON INSTRUMENT CO
PI MARKER 9.50 200UL
Supplier: PROTEIN SIMPLE MS
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Western Blot Boxes
Supplier: Alkali Scientific
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS
Pierce™ Surfact-Amps™ Detergent Solutions, Thermo Scientific
Supplier: Invitrogen