STAIN HISTOCORE SPECTRA H/E SYS S1-800
MSPP-3801656
Frequently Bought Together
VWR® Razor Blades
: VWR International
Corning® Centrifuge Tubes, 15 ml
: Corning
VWR® Disposable Serological Pipettes
: VWR International
Alconox® Powdered Precision Cleaners
: Alconox
VWR® Broken Glass Disposal Cartons
: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
: VWR International
VWR® Disposable Petri Dishes
: VWR International
InVitro™ Biotainer® Carboys, PETG, Sterile, Thermo Scientific
: Thermo Fisher Scientific
VWR® Sterile 70% Isopropanol
: VWR International
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
: Hardy Diagnostics