ANTIBODY CD159A PE-VIO-770 100T 200UL
MSPP-130113567
Frequently Bought Together
DEFIBRINATED BLOOD SHEEP 100ML
: LAMPIRE BIOLOGICAL LAB MS
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
: Invitrogen
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
: MABTECH INC MS
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
Bovine Plasma with Citrate, Equitech- Bio, Inc.
: Equitech-Bio
74042 PHORBOL12 MYRISTATE 13 ACETATE 1MG
: STEMCELL Technologies
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
: Innovative Research Inc
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
: Meso Scale Diagnostics
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
: Invitrogen
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
: INDIGO BIOSCIENCES INC MS
BUFFER STABILIZR 7.5% DTPA-PURI BSA 50ML
: Revvity Health Sciences
73254 METFORMIN HYDROCHLORIDE 5G
: STEMCELL Technologies
72262 ALL TRANS RETINOIC ACID 50MG
: STEMCELL Technologies
SOLUTION ATTUNE WASH 250ML
: Invitrogen
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
: Invitrogen