WIZARD SCREEN HT DEEP WL BLOCK FORMT 1ML
Catalog # MSPP-HR2-463
Supplier: HAMPTON RESEARCH MS
WIZARD SCREEN HT DEEP WL BLOCK FORMT 1ML
Catalog # MSPP-HR2-463
Supplier: HAMPTON RESEARCH MS
Supplier Number: HR2-463
Frequently Bought Together
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×5 Configuration
Supplier: Alkali Scientific
ProSieve™ Protein Markers
Supplier: Lonza
CASTING TRAY FOR ELECTROPHORESIS SYSTEM
Supplier: G-Biosciences
AGAROSE ALFA SELECTOR ST MAGNETIC 2ML
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS
MORPHEUS HT96
Supplier: MOLECULAR DIMENSIONS INC MS
Buffer P1 (Red), Zymo Research
Supplier: Zymo Research
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
FLUOTAG X2 ANTI ALFA ALEXAFLUOR 647 2ML
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS
Protein Transfer Buffers, G-Biosciences
Supplier: G-Biosciences
GelBond® Film
Supplier: Lonza
FACS and IF Staining Buffer with BSA
Supplier: Rockland Immunochemical
Incubation Trays, Azure Biosystems
Supplier: Azure Biosystems
BLOK™ Casein, 1% Casein Protein Blocking Solution, G-Biosciences
Supplier: G-Biosciences
Iodoacetamide
Supplier: G-Biosciences
picoLUCENT™ PLUS ECL Reagent for Western blotting, G-Biosciences
Supplier: G-Biosciences
Pierce™ Ni-NTA Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
CRYSTAL SCREEN MCSG-1 1.7 ML BLOCK
Supplier: 0000032292