BUFFER MOPS SDS RUNNING 20X 1L
Catalog # MSPP-105300072
Supplier: 0000014548
BUFFER MOPS SDS RUNNING 20X 1L
Catalog # MSPP-105300072
Supplier: 0000014548
Supplier Number: 10530007-2
Frequently Bought Together
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Hydrogen peroxide (30 - <lt/> 40%) 30% stabilised ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
TAE 50X Buffer, MP Biomedicals
Supplier: MP Biomedicals
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Western Blot Boxes
Supplier: Alkali Scientific
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research