BUFFER LAEMMLI SDS SAMP 6X NON REDUCING2
Catalog # MSPP-105700191
Supplier: 0000014548
BUFFER LAEMMLI SDS SAMP 6X NON REDUCING2
Catalog # MSPP-105700191
Supplier: 0000014548
Supplier Number: 10570019-1
Frequently Bought Together
FOCUS DTT 2M
Supplier: 0000014548
BUFFER A.C.E. PHOSPHATE 50MM PH 6.5
Supplier: 0000014548
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
DNA Elution Buffer, Zymo Research
Supplier: Zymo Research
Electrophoresis Running Buffers, G-Biosciences
Supplier: G-Biosciences
Upright Stainless Steel Freezer Rack for 2" Boxes, 4×6 Configuration
Supplier: Alkali Scientific
Buffer P3 (Yellow), Zymo Research
Supplier: Zymo Research
Gelatin
Supplier: G-Biosciences
Pierce™ Protein A/G Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
Endo-Wash Buffer, Zymo Research
Supplier: Zymo Research
1X TBS pH 7.8
Supplier: Rockland Immunochemical
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
N,N,N',N'-Tetramethylethylenediamine (TEMED) for electrophoresis
Supplier: Thermo Scientific Chemicals
GelBond® Film
Supplier: Lonza
Stainless Steel 3×3 Upright Drawer Rack for 2" Boxes
Supplier: Alkali Scientific
Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
Tube-O-DIALYZER™ Micro Dialysis System, G-Biosciences
Supplier: G-Biosciences
ScanLater™ Western Blot Assay Kit, Molecular Devices
Supplier: Molecular Devices
2D Electrophoresis Reagents, G-Biosciences
Supplier: G-Biosciences