Order Entry
United States
Orders LinkContactUsLinkComponent
Hydrometer Racks

Hydrometer Racks

Hydrometer racks, polypropylene, gray, 11.6×6.9×9.2"
Catalog # 77591-822
Supplier:   Heathrow Scientific
Supplier #:  121002
Hydrometer Racks
Catalog # 77591-822
Hydrometer Racks
Hydrometer Racks

Hydrometer racks, polypropylene, gray, 11.6×6.9×9.2"

Price
$88.10
New Customer Promotional Price
Log in for availability
Each Case of 6
Product Details & Documents

These hydrometer racks accomdate up to 10 hydrometers with diameters up to 31 mm, ensuring safe storage, efficient drying, and secure transport.

  • Teardrop-shaped slots hold hydrometers securely in place, support a range of diameters, and ensure easy insertion/removal for convenient handling
  • 3-Tier setup provides enhanced stability and protection of hydrometer body
  • Built-in handles provide a secure grip for safe and convenient transport
  • Removable drip tray simplifies cleaning and maintenance, improving sanitation and protecting your workspace
  • Stable wide base ensures increased stability and secure placement for hydrometers

Durable polypropylene construction is easy to clean, long-lasting, and fully recyclable. Stable wide base ensures increased stability and secure placement for hydrometers.

Specifications
  • Description:
    Hydrometer racks, polypropylene, gray
  • Overall dimensions:
    11.6×6.9×9.2"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.