Order Entry
United States
Orders LinkContactUsLinkComponent
Cell Culture Inserts, flexiPERM®

Cell Culture Inserts, flexiPERM®

flexiPERM® cell culture insert, slide, 8, 0.9 cm²
Catalog # 77589-814
Supplier:   SARSTEDT INC
Supplier #:  94.6032.039
Cell Culture Inserts, flexiPERM®
Catalog # 77589-814
Supplier:  SARSTEDT INC
Cell Culture Inserts, flexiPERM®
Cell Culture Inserts, flexiPERM®

flexiPERM® cell culture insert, slide, 8, 0.9 cm²

Price
$431.33
Log in for availability
Pack of 5 Case of 100
Product Details & Documents

To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.

  • Material: silicone
  • Purity: Non-pyrogenic/endotoxin-free, non-cytotoxic
  • Reusable silicone adhesive cell culture inserts to divide cell culture vessels and slides into smaller units
  • Can be used for approx. 2 weeks in long-term tests
  • Sterilizable with autoclave or 70% ethanol
  • Heat-resistant (up to 125 °C), cold-resistant (up to -20 °C), high chemical resistance
  • Five different formats: flexiPERM® slide and micro12 for parallel tests on slides, flexiPERM® conA and conB for micromanipulation/microinjection, flexiPERM® disc for use in cell culture dishes e.g. for co-cultivation

SARSTEDT flexiPERM® reusable silicone inserts subdivide slides and any cell culture dish into smaller cultivation units. Different cell lines can be grown in one vessel or equivalent cell lines can be grown in parallel with the assurance of comparable growth conditions. flexiPERM® inserts are highly adhesive to any flat surface for up to 14 days without additional or potentially contaminating adhesives. flexiPERM® inserts are easily removed after use without tools and may be sterilized and reused at least 10 times.

Specifications
  • No. of wells:
    8
  • Description:
    flexiPERM® cell culture insert, slide
  • Growth area:
    0.9 cm²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.