Order Entry
United States
Orders LinkContactUsLinkComponent
 

Feces Tubes

Feces tubes, PP, Non sterile, Transparent, 25 mm, 107 mm, BAGA
  77589-120
 :  80.622.111
Feces Tubes
  77589-120
Feces Tubes
Feces Tubes

Feces tubes, PP, Non sterile, Transparent, 25 mm, 107 mm, BAGA

 
$275.13
   
Case of 500
 

To meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, Sarstedt offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.

  • Material: PP
  • Particularly liquid and odor-tight
  • Larger opening to better collect the sample
  • Secure screw cap for sending samples
  • Stands firmly due to skirted base
  • Options with paper labels and white material for light protection

To hygienically collect stool samples, all collection vessels are fitted with integral spoons. Fecal collection tubes combined with a broad and sturdy scoop in the screw cap make sampling easy.

 
  • Ø:
    25 mm
  • Sterility:
    Non sterile
  • Color:
    Transparent
  • Description:
    Feces tubes
  • Length:
    107 mm
  • Material:
    PP
  • Packaging:
    BAGA