Order Entry
United States
Orders LinkContactUsLinkComponent
Feces Tubes with Flat Base and Push Cap

Feces Tubes with Flat Base and Push Cap

PS, Non sterile, 23.5 mm, 75 mm, BAGA
Catalog # 77589-114
Supplier:   SARSTEDT INC
Supplier #:  80.620
Feces Tubes with Flat Base and Push Cap
Catalog # 77589-114
Supplier:  SARSTEDT INC
Feces Tubes with Flat Base and Push Cap
Feces Tubes with Flat Base and Push Cap

PS, Non sterile, 23.5 mm, 75 mm, BAGA

Price
$227.19
Log in for availability
Case of 500
Product Details & Documents

In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, this offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.

  • Material: PS
  • Particularly liquid and odor-tight
  • Larger opening to better collect the sample
  • Stands firmly due to skirted base

To hygienically collect stool samples, all collection vessels are fitted with integral spoons. Fecal collection tubes combined with a broad and sturdy scoop in the screw cap make sampling easy.

Specifications
  • Ø:
    23.5 mm
  • Sterility:
    Non sterile
  • Length:
    75 mm
  • Material:
    PS
  • Packaging:
    BAGA
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.