Order Entry
United States
Orders LinkContactUsLinkComponent
 

Absorbent Liners for Transport Containers

Absorbent liners for mailing container 84×30 mm, 8 ml suction capacity, 75×65 mm, CS
Catalog # 77589-104
Supplier:   SARSTEDT INC
 :  78.684
Absorbent Liners for Transport Containers
Catalog # 77589-104
Supplier:  SARSTEDT INC
Absorbent Liners for Transport Containers
Absorbent Liners for Transport Containers

Absorbent liners for mailing container 84×30 mm, 8 ml suction capacity, 75×65 mm, CS

Price
$295.75
Log in for availability
Case of 2,000
Product Details & Documents

SARSTEDT offers sample transport systems and secondary containers for a wide variety of transport requirements.

  • Material: Cellulose
  • Separate absorbent liners suitable for various mailing containers
  • Various designs for different absorption capacities

For use with mailing containers with screw cap; a secure container for blood, urine and stool sample tubes.

Specifications
  • Description:
    Absorbent liners for mailing container 84×30 mm, 8 ml suction capacity
  • Dimensions:
    75×65 mm
  • Packaging:
    CS
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.