Order Entry
United States
Orders LinkContactUsLinkComponent
Absorbent Liners for Transport Containers

Absorbent Liners for Transport Containers

Absorbent liners for mailing container 126ר 30 mm, 12 ml suction capacity, 75×90 mm, CS
Catalog # 77589-100
Supplier:   SARSTEDT INC
Supplier #:  78.670
Absorbent Liners for Transport Containers
Catalog # 77589-100
Supplier:  SARSTEDT INC
Absorbent Liners for Transport Containers
Absorbent Liners for Transport Containers

Absorbent liners for mailing container 126ר 30 mm, 12 ml suction capacity, 75×90 mm, CS

Price
$178.04
Log in for availability
Pack of 1,000 Case of 4000
Product Details & Documents

SARSTEDT offers sample transport systems and secondary containers for a wide variety of transport requirements.

  • Material: Cellulose
  • Separate absorbent liners suitable for various mailing containers
  • Various designs for different absorption capacities

For use with mailing containers with screw cap; a secure container for blood, urine and stool sample tubes.

Specifications
  • Description:
    Absorbent liners for mailing container 126ר 30 mm, 12 ml suction capacity
  • Dimensions:
    75×90 mm
  • Packaging:
    CS
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.