Screw Cap Microtubes
Screw cap microtubes with skirted conical base and knurl, Amber, 0.5 ml, —, —, BAGA, Non sterile
77588-962
: 72.730.304
SARSTEDT screw cap microtubes provide an exceptionally secure seal with the integrated O-ring. Ideal for centrifugation, transport and storage of high-quality oligonucleotides, enzymes and buffers.
- Material: PP
- Robust polypropylene material for safety and versatility
- 3 different volumes meet filling requirements from a few microliters to 2 milliliters
- Leak-resistant according to ADR and IATA- tested at a vacuum of 0.95 bar
- Centrifugation up to 20000×g
- Can be autoclaved to 121 °C and frozen to –80 °C
- O-ring cap increases cap security (IATA tested)
With volumes from 0.5 to 2.0 ml, a variety of container styles and caps, plus a wide selection of coding options, SARSTEDT screw cap micro tubes offer the right solution for your every requirement.
- Cap type:—
- Sterility:Non sterile
- Volume:0.5 ml
- Color:Amber
- Description:Screw cap microtubes with skirted conical base and knurl
- Packaging:BAGA
- Cap color:—



