Order Entry
United States
Orders LinkContactUsLinkComponent
 

Feces Tubes

Feces tubes, with paper label, PP, Non sterile, Transparent, 20 mm, 76 mm, BAGA
Catalog # 77589-144
Supplier:   SARSTEDT INC
 :  80.734.301
Feces Tubes
Catalog # 77589-144
Supplier:  SARSTEDT INC
Feces Tubes
Feces Tubes

Feces tubes, with paper label, PP, Non sterile, Transparent, 20 mm, 76 mm, BAGA

Price
$254.62
Log in for availability
Case of 500
Product Details & Documents

To meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, Sarstedt offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.

  • Material: PP
  • Particularly liquid and odor-tight
  • Larger opening to better collect the sample
  • Secure screw cap for sending samples
  • Stands firmly due to skirted base
  • Options with paper labels and white material for light protection

To hygienically collect stool samples, all collection vessels are fitted with integral spoons. Fecal collection tubes combined with a broad and sturdy scoop in the screw cap make sampling easy.

Specifications
  • Ø:
    20 mm
  • Sterility:
    Non sterile
  • Color:
    Transparent
  • Description:
    Feces tubes, with paper label
  • Length:
    76 mm
  • Material:
    PP
  • Packaging:
    BAGA