Low Retention Pipette Tips
Product Details & Documents
Proven, high-quality pipette tips provide maximum precision, sophisticated design, and the highest purity. By using these low retention tips, costly and sensitive reagents can be more efficiently transferred with less loss, improving test results.
One system with four ordering options to fit every requirement: Box, SingleRefill, StackPack and bag
SingleRefill uses 41% less and StackPack uses 64% less material than continually ordering a regular box system
Reduce waste; after use the individual components of the box system can fit into each other, saving disposal volume and reducing waste
Volume, batch, and shelf life are laser coated on each tray for easy identification
The box is a universal and robust solution at the heart of this new pipette tip system.
The SingleRefill is an easy way to refill filter tips and eliminate unnecessary waste by reusing the existing box. The closed design prevents contamination of the pipette tips, meaning the refill process is hygienic as well as environmentally friendly.
The StackPack accommodates higher sample processing rates that require more efficient solutions. Individual trays can be transferred to the box in one simple maneuver, plus the compact and stable design means that tips can be removed for use directly from the StackPack itself.
The bag provides a low-cost packaging solution, which can be hygienically opened and sealed again using the pressure seal. Additional tamper-evident seal also gives users an extra reassurance that tips are 100% free from contamination.
Specifications
- Volume:10 µl
- Length:31 mm
- Tip color:Natural
- Packed:96 tips/box, 20 boxes
- Filter:+
- Sterile:+
- Tip style:Low retention
- Color code:Grey
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



