Order Entry
United States
Orders LinkContactUsLinkComponent
Micro Needles

Micro Needles

Micro needle 23G × ¾", blue, 36 mm, Sterile, non-pyrogenic/endotoxin-free
Catalog # 77589-492
Supplier:   SARSTEDT INC
Supplier #:  85.923
Micro Needles
Catalog # 77589-492
Supplier:  SARSTEDT INC
Micro Needles
Micro Needles

Micro needle 23G × ¾", blue, 36 mm, Sterile, non-pyrogenic/endotoxin-free

Price
$147.87
Log in for availability
Pack of 50 Case of 500
Product Details & Documents

The micro needles were developed to meet the specific needs of neonatology. It has a grooved holder to ensure safe puncture and enables turning the needle in the vein for better positioning. The short design of the needle means a minimal dead volume during sample collection.

  • Ideal for venous blood collection of even the tiniest quantities
  • Minimal blood loss with no residual volume in the holder
  • Ridged holder for a secure grip allows optimal handling of the needle
  • Can be rotated 360° in difficult vein conditions
  • Exceptional needle quality and exposed outlet opening enable free blood flow

These pediatric blood collection products make the process of collecting blood from your smallest patients as gentle and painless as possible. The priority is minimizing the fill volume of each collection and keeping the diagnostic blood loss as low as possible.

Documents

Specifications
  • Sterility:
    Sterile, non-pyrogenic/endotoxin-free
  • Description:
    Micro needle 23G × ¾", blue
  • Length:
    36 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.