Safety Lancets
Safety lancet, penetration depth: 1.6 mm, 28 G, Light blue
Catalog # 77589-418
Supplier: SARSTEDT INC
Supplier #: 85.1015
Jump to:
Product Details & Documents
Sarstedt safety lancets guarantee optimal safety for patients and users and are flexible with regard to penetration depth and required quantity of blood. The needle and blade are always safely concealed in the lancet body before and after use, which prevents needlestick injuries and cross-contamination.
- Safety for users and patients
- Sterile, single-use system
- Patient-friendly handling activated by a release button
- Large selection of penetration depths and widths depending on the quantity of blood and skin conditions
Sarstedt's pre-analytical product range includes products for optimal specimen collection. The capillary collection systems are among the most versatile on the market.
Specifications
- Color:Light blue
- Description:Safety lancet, penetration depth: 1.6 mm
- Gauge:28 G