Screw Cap Microtubes
Screw cap microtubes with assembled closure,with skirted conical base and knurl with printed label, Transparent, 0.5 ml, —, Retaining loop cap, BAGA, Sterile
Catalog # 77588-952
Supplier: SARSTEDT INC
Supplier #: 72.730.106
Jump to:
Product Details & Documents
SARSTEDT screw cap microtubes provide an exceptionally secure seal with the integrated O-ring. Ideal for centrifugation, transport and storage of high-quality oligonucleotides, enzymes and buffers.
- Material: PP
- Robust polypropylene material for safety and versatility
- 3 different volumes meet filling requirements from a few microliters to 2 milliliters
- Leak-resistant according to ADR and IATA- tested at a vacuum of 0.95 bar
- Centrifugation up to 20000×g
- Can be autoclaved to 121 °C and frozen to –80 °C
- O-ring cap increases cap security (IATA tested)
With volumes from 0.5 to 2.0 ml, a variety of container styles and caps, plus a wide selection of coding options, SARSTEDT screw cap micro tubes offer the right solution for your every requirement.
Documents
Specifications
- Cap type:Retaining loop cap
- Sterility:Sterile
- Volume:0.5 ml
- Color:Transparent
- Description:Screw cap microtubes with assembled closure,with skirted conical base and knurl with printed label
- Packaging:BAGA
- Cap color:—
Accessories
Product Information
- Catalog #PackagingAvailabilityPrice
- Decapper for screw cap micro tubes, aluminium, sterile
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



