Orders LinkContactUsLinkComponent
Dual Position Ventilation Caps
Dual Position Ventilation Caps
Dual position closure cap, LDPE, 16 mm, Neutral
Catalog # 77588-232
Supplier:   SARSTEDT INC
Supplier #:  65.725
Dual Position Ventilation Caps
Catalog # 77588-232
Supplier:  SARSTEDT INC
Dual Position Ventilation Caps
Dual Position Ventilation Caps
Dual position closure cap, LDPE, 16 mm, Neutral
Price
$6.89
Log in for availability
Pack of 100 Case of 5000
Product Details & Documents
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Two position fit allows both gas exchange and sealing
  • Ribbed surface for maximum grip
Dual position ventilation caps offer excellent sealing properties, and are available in LDPE material.
Specifications
  • Description:
    Dual position closure cap
  • Material:
    LDPE
  • For tube O.D.:
    16 mm
  • Cap color:
    Neutral