Forensics Smear Swabs
Forensics smear swab, with ventilation membrane, PS, 85 mm, Viscose
Catalog # 77589-136
Supplier: SARSTEDT INC
Supplier #: 80.630
Jump to:
Product Details & Documents
SARSTDT forensic swabs contain a ventilation membrane in the tube base, allowing the swab to dry in the tube.
- Round swab shape
- Complies with all requirements of ISO 18385
- Employee DNA exclusion database available
An EtO gassing process developed specifically to address the requirements of forensics, as well as the highest cleanroom conditions in the production process, guarantee that these forensic swabs are absolutely free from DNA.
Specifications
- Description:Forensics smear swab, with ventilation membrane
- Handle length:85 mm
- Handle material:PS
- Head material:Viscose