Order Entry
United States
Orders LinkContactUsLinkComponent
Cuvettes

Cuvettes

Cuvettes, Rack pack, PMMA
Catalog # 77588-356
Supplier:   SARSTEDT INC
Supplier #:  67.738
Cuvettes
Catalog # 77588-356
Supplier:  SARSTEDT INC
Cuvettes
Cuvettes

Cuvettes, Rack pack, PMMA

Price
$28.07
Log in for availability
Pack of 100 Case of 2000
Product Details & Documents

SARSTEDT has been known as a manufacturer of high-quality disposable cuvettes.

  • Height: 45 mm
  • Optical sides: 2
  • Path length: 10 mm
  • Disposable cuvette made of high-quality, crystal-clear polystyrene or acrylic (PMMA)
  • Ideal for photometric analyses of liquids in the wavelength range ≥300 nm (PMMA)/≥330 nm (PS)
  • An arrow integrated in the cuvette allows exact positioning in the photometer
  • Nest sorting ensures consistent transmission values within a polystyrene box

SARSTEDT cuvettes are made from polystyrene (PS), acrylic (PMMA) and a special plastic for analysis in the UV range, which are ideal for photometric analysis of liquid solutions. The cuvettes are also available in 2× optical semi-micro and a 4× optical format for 90° angle fluorescence measurements.

Specifications
  • Description:
    Cuvettes
  • Material:
    PMMA
  • Packaging:
    Rack pack