Color-Coded Inserts for CryoPure Tubes
Blue
Catalog # 77588-114
Supplier: SARSTEDT INC
Supplier #: 65.386.006
Jump to:
Product Details & Documents
High-quality, safe and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Polystyrene material
- Suitable for use with CryoPure tubes
- Visual coding for easy identification of samples
- Colors available individually or as a color mix
Up to 36 color combinations are available in combination with the screw caps in 6 different colors. Assorted color options include a mix of White, Red, Yellow, Green, and Purple.
Specifications
- Color:Blue