Order Entry
United States
Orders LinkContactUsLinkComponent
Tubes without Cap, Round Base, PS

Tubes without Cap, Round Base, PS

Without label/print, 5 ml, 13 mm, 75 mm, Non graduated, BAGA
Catalog # 77587-550
Supplier:   SARSTEDT INC
Supplier #:  55.475.305
Tubes without Cap, Round Base, PS
Catalog # 77587-550
Supplier:  SARSTEDT INC
Tubes without Cap, Round Base, PS
Tubes without Cap, Round Base, PS

Without label/print, 5 ml, 13 mm, 75 mm, Non graduated, BAGA

Price
$57.88
Log in for availability
Pack of 1,000 Case of 2000
Product Details & Documents

High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, this range offers the right product for every laboratory and analytical requirement.

  • Material: PS
  • Base shape: Round base
  • Amount of packaging reduced by 40% (if StackPack option is used)
  • Polystyrene (PS) tubes for maximum transparency
  • Different push caps for a variety of applications
  • Volumes from 3.5 to 26 ml

This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.

Specifications
  • Ø:
    13 mm
  • Volume:
    5 ml
  • Description:
    Without label/print
  • Length:
    75 mm
  • Packaging:
    BAGA
  • Graduations:
    Non graduated