Push Caps
LDPE, Tube Ø 12 mm, Purple, BAGA
77588-330
: 65.809.303
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- LDPE material
- Large grip for optimal handling
- Quick opening and closing of tubes
- Ribbed surface for maximum grip
Push caps offer excellent sealing properties, and are available in a wide variety of colors to facilitate organization and coding.
- Material:LDPE
- Packaging:BAGA
- Fits:Tube Ø 12 mm
- Cap color:Purple