Push Caps
LDPE, Tube Ø 11.5 and 12 mm, Neutral, BAGA
Catalog # 77588-322
Supplier: SARSTEDT INC
: 65.809.021
Jump to:
Product Details & Documents
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- LDPE material
- Large grip for optimal handling
- Quick opening and closing of tubes
- Ribbed surface for maximum grip
Push caps offer excellent sealing properties, and are available in a wide variety of colors to facilitate organization and coding.
Specifications
- Material:LDPE
- Packaging:BAGA
- Fits:Tube Ø 11.5 and 12 mm
- Cap color:Neutral